Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583854
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583854 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583854 Supplier Invitrogen™ Supplier No. PA583854
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EITEEMMDQA NDKKVAAIEA LNDGELQKAI DLFTDAIKLN PRLAILYAKR ASVFVKLQKP NAAIRDCDRA Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 93%.

The protein encoded by this gene is an adaptor protein that mediates the association of the heat shock proteins HSP70 and HSP90. This protein has been shown to be involved in the assembly process of glucocorticoid receptor, which requires the assistance of multiple molecular chaperones. The expression of this gene is reported to be downregulated in colorectal carcinoma tissue suggesting that is a candidate tumor suppressor gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene St13
Gene Accession No. P50502
Gene Alias 1110007I03Rik; 3110002K08Rik; AAG2; aging-associated protein 2; AW555194; FAM10A1; FAM10A4; heat shock 70kD protein binding protein; hedgehog interacting protein; hedgehog-interacting protein; HHIP; HIP; HOP; Hsc70-interacting protein; Hsp70 interacting protein; Hsp70-interacting protein; HSPABP; HSPABP1; hypothetical protein; p48; PRO0786; Progesterone receptor-associated p48 protein; Protein FAM10A1; Protein ST13 homolog; putative tumor suppressor ST13; RCJMB04_6h13; Renal carcinoma antigen NY-REN-33; SNC6; ST13; ST13 Hsp70 interacting protein; ST13, Hsp70 interacting protein; ST13P5; suppression of tumorigenicity 13; suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein); suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein) pseudogene 5; suppression of tumorigenicity 13 (colon carcinoma) Hsp70-interacting protein; suppression of tumorigenicity 13 protein; testis secretory sperm-binding protein Li 233m; UNQ5825/PRO19644
Gene Symbols St13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ST13. Recombinant protein control fragment (Product #RP-99511).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6767
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.