Learn More
HIP1/Huntingtin-interacting Protein 1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305176A

Description
The recommended shelf life for this product is 1 year from date of receipt.
The product of this gene is a membrane-associated protein that colocalizes with huntingtin. This protein has similarities to cytoskeleton proteins and its interaction with huntingtin is thought to play a functional role in the cell filament network. Loss of normal huntingtin-HIP1 interaction in Huntington disease may contribute to a defect in membrane-cytoskeletal integrity in the brain. This gene could help in the understanding of the normal function of huntingtin and also the pathogenesis of Huntington disease. It also has been implicated in the pathogenesis of hematopoietic maligcies. An alternative splice variant of this gene has been described but its full length sequence has not been determined.Specifications
HIP1/Huntingtin-interacting Protein 1 | |
Polyclonal | |
Unconjugated | |
HIP1 | |
HIP I, Huntingtin interacting protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3092 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
O00291 | |
HIP1 | |
Between 987 and 1037 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.