Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HIP1 Monoclonal Antibody (1B11)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA116748
Description
This antibody does not react with rat samples. Suggested positive control: Hela whole cell extract, HeLa whole cell lysate.
Huntingtin disease, a neurodegenerate disease, is caused by the expansion of a polymorphic glutamine tract in huntingtin. The Huntingtin Interacting Protein 1 (HIP-1) is a reportedly proapoptotic, cargo-specific adaptor protein that may be involved in the pathogenesis of Huntingtin disease. As well as playing a role in Huntingtin disease, it is likely to be involved in the recruitment of clathrin coats to lipid membranes and it may also factor in tumorigenesis by allowing the survival of precancerous and cancerous cells. Since HIP-1 expression is significantly associated with prostate and colon cancer metastasis, HIP-1 can serve as a putative prognostic factor for prostate and colon cancers.
Specifications
HIP1 | |
Monoclonal | |
Conc. Not Determined | |
Ascites with 0.1% sodium azide | |
O00291, Q8VD75 | |
HIP1 | |
Human HIP1 (~65 kDa). | |
RUO | |
215114, 3092 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunoprecipitation, Western Blot, Immunocytochemistry | |
1B11 | |
Unconjugated | |
HIP1 | |
2610109B09Rik; A930014B11Rik; E130315I21Rik; HIP I; hip1; HIP-1; HIP-I; huntingtin interacting protein 1; huntingtin-interacting protein 1; Huntingtin-interacting protein I; ILWEQ; Kiaa4113; mKIAA4113; SHON; SHONbeta; SHONgamma | |
Mouse | |
100 μL | |
Primary | |
Human, Mouse, Bacteria | |
Antibody | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction