Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA593288
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA593288 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA593288 Supplier Invitrogen™ Supplier No. PA593288
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SELEADLAEQ QHLRQQAADD CEFLRAELDE LRRQREDTEK AQRSLSEIER KAQANEQRYS KLKEKYSELV QNHADLLRKN AEVTKQVSMA RQAQVDLERE KKELEDSLER ISDQGQRKTQ EQLEVLESLK QELATSQREL QVLQGSLETS AQSEANWAAE FAELEKERDS LVSGAAHREE ELSALRKELQ DTQLKLASTE ESMCQLAKDQ RKMLLVGSRK AAEQVIQDAL NQLEEPPLIS CAGSADHLLS TVTSISSCIE QLEKSWSQYL ACPEDISGLL H; Positive Samples: A-549, BT-474, SW480, 293T, Mouse brain; Cellular Location: Cytoplasm, Cytoplasmic vesicle, Endomembrane system, Nucleus, clathrin-coated vesicle membrane.

Huntingtin disease, a neurodegenerate disease, is caused by the expansion of a polymorphic glutamine tract in huntingtin. The Huntingtin Interacting Protein 1 (HIP-1) is a reportedly proapoptotic, cargo-specific adaptor protein that may be involved in the pathogenesis of Huntingtin disease. As well as playing a role in Huntingtin disease, it is likely to be involved in the recruitment of clathrin coats to lipid membranes and it may also factor in tumorigenesis by allowing the survival of precancerous and cancerous cells. Since HIP-1 expression is significantly associated with prostate and colon cancer metastasis, HIP-1 can serve as a putative prognostic factor for prostate and colon cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIP1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HIP1
Gene Accession No. O00291, Q8VD75
Gene Alias 2610109B09Rik; A930014B11Rik; E130315I21Rik; HIP I; hip1; HIP-1; HIP-I; huntingtin interacting protein 1; huntingtin-interacting protein 1; Huntingtin-interacting protein I; ILWEQ; Kiaa4113; mKIAA4113; SHON; SHONbeta; SHONgamma
Gene Symbols HIP1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 410-690 of human HIP1 (NP_0053293).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 215114, 3092
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.