Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIP1 Related Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | HIP1 Related |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
HIP1 Related Polyclonal specifically detects HIP1 Related in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
HIP1 Related | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
FLJ14000, HIP-12, HIP12HIP1-related protein, HIP3, huntingtin interacting protein 1 related, huntingtin interacting protein 12, Huntingtin-interacting protein 12, huntingtin-interacting protein 1-related protein, ILWEQ, KIAA0655FLJ27022, MGC47513 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 300-600 of human HIP1 Related (NP_003950.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, Cell Biology, Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
PBS with 50% glycerol, pH7.3. | |
9026 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title