Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIP1R Monoclonal Antibody (1E1)
GREENER_CHOICE

Catalog No. AMPA6750
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AMPA6750 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AMPA6750 Supplier Invitrogen™ Supplier No. MA116750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Suggested positive control: HeLa whole cell lysate and rat brain homogenate, Hela whole cell lysate, antigen standard for IFT57 (transient overexpression lysate), rat brain protein.

Huntingtin disease, a neurodegenerate disease, is caused by the expansion of a polymorphic glutamine tract in huntingtin. The Huntingtin Interacting Protein 1 (HIP-1) is a reportedly proapoptotic, cargo-specific adaptor protein that may be involved in the pathogenesis of Huntingtin disease. As well as playing a role in Huntingtin disease, it is likely to be involved in the recruitment of clathrin coats to lipid membranes and it may also factor in tumorigenesis by allowing the survival of precancerous and cancerous cells. Since HIP-1 expression is significantly associated with prostate and colon cancer metastasis, HIP-1 can serve as a putative prognostic factor for prostate and colon cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIP1R
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1E1
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.1% sodium azide
Gene HIP1R
Gene Accession No. O75146, Q9JKY5
Gene Alias AA410023; HIP12; HIP-12; Hip1r; HIP1-related protein; HIP3; huntingtin interacting protein 1 related; huntingtin interacting protein 12; Huntingtin-interacting protein 12; huntingtin-interacting protein 1-related protein; ILWEQ; KIAA0655; mKIAA0655
Gene Symbols HIP1R
Host Species Mouse
Immunogen Human HIP1 Related-TALIN Homology Region (57 kDa).
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29816, 81917, 9026
Target Species Human, Mouse, Rat, Bacteria
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.