Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583038
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583038 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583038 Supplier Invitrogen™ Supplier No. PA583038
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VTGAICLDIL KDQWAAAMTL RTVLLSLQAL LAAAEPDDPQ DAVVANQYKQ NPEMFKQTAR LWAHVYAGAP VSSPEYTKKI ENLCAMGFDR NAVIVALSSK SWDVETATEL Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.

Accepts ubiquitin from the E1 complex and catalyzes its covalent attachment to other proteins. In vitro, in the presence or in the absence of BRCA1-BARD1 E3 ubiquitin-protein ligase complex, catalyzes the synthesis of 'Lys-48'-linked polyubiquitin chains. Does not transfer ubiquitin directly to but elongates monoubiquitinated substrate protein. Mediates the selective degradation of short-lived and abnormal proteins, such as the endoplasmic reticulum-associated degradation (ERAD) of misfolded lumenal proteins. Ubiquitinates huntingtin. May mediate foam cell formation by the suppression of apoptosis of lipid-bearing macrophages through ubiquitination and subsequence degradation of p53/TP53. Proposed to be involved in ubiquitination and proteolytic processing of NF-kappa-B; in vitro supports ubiquitination of NFKB1. In case of infection by cytomegaloviruses may be involved in the US11-dependent degradation of MHC class I heavy chains following their export from the ER to the cytosol. In case of viral infections may be involved in the HPV E7 protein-dependent degradation of RB1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIP2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UBE2K
Gene Accession No. P61086
Gene Alias AW492011; D5Ertd601e; DKFZp564C1216; DKFZp686J24237; E2 ubiquitin-conjugating enzyme K; E2(25K); E2-25k; HIP2; HIP-2; huntingtin interacting protein 2; huntingtin-interacting protein 2; HYPG; LIG; UBC1; UBE2K; Ubiquitin carrier protein; ubiquitin conjugating enzyme E2 K; ubiquitin conjugating enzyme E2K; ubiquitin-conjugating enzyme E2 K; Ubiquitin-conjugating enzyme E2(25K); ubiquitin-conjugating enzyme E2-25 KDA; ubiquitin-conjugating enzyme E2-25K; ubiquitin-conjugating enzyme E2K; ubiquitin-conjugating enzyme E2K (UBC1 homolog, yeast); ubiquitin-protein ligase
Gene Symbols UBE2K
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UBE2K. Recombinant protein control fragment (Product #RP-102065).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3093
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.