Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIP55 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582717
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582717 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582717 Supplier Invitrogen™ Supplier No. PA582717
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: INWTGEGVND VRKGACASHV STMASFLKGA HVTINARAEE DVEPECIMEK VAKASGANYS FHKESGRFQD VGPQAPVGSV YQ Highest antigen sequence indentity to the following orthologs: Mouse - 95%, Rat - 95%.

The protein encoded by this gene is one of the highly conserved mini-chromosome maintenance proteins (MCM) that are essential for the initiation of eukaryotic genome replication. The hexameric protein complex formed by MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. The MCM complex consisting of this protein and MCM2, 6 and 7 proteins possesses DNA helicase activity, and may act as a DNA unwinding enzyme. The phosphorylation of this protein by CDC2 kinase reduces the DNA helicase activity and chromatin binding of the MCM complex. This gene is mapped to a region on the chromosome 8 head-to-head next to the PRKDC/DNA-PK, a DNA-activated protein kinase involved in the repair of DNA double-strand breaks. Alternatively spliced transcript variants encoding the same protein have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIP55
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DBNL
Gene Accession No. Q62418, Q9JHL4, Q9UJU6
Gene Alias Abp1; Actin-binding protein 1; cervical mucin-associated protein; Cervical SH3P7; CMAP; DBNL; dbnl {ECO:0000312; drebrin F; drebrin like; drebrin-F; drebrin-like; drebrin-like protein; EMBL:AAH72483.1}; HIP55; HIP-55; HPK1-interacting protein of 55 kDa; mAbp1; PP5423; SH3 domain-containing protein 7; Sh3p7; src homology 3 domain-containing protein HIP-55
Gene Symbols DBNL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DBNL. Recombinant protein control fragment (Product #RP-92735).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13169, 28988, 83527
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.