Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIPK1/HIPK2/HIPK3 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5106247
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106247 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106247 Supplier Thermo Scientific Supplier No. PA5106247

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total HIPK1/2/3.

The protein encoded by this gene belongs to the Ser/Thr family of protein kinases and HIPK subfamily. It phosphorylates homeodomain transcription factors and may also function as a co-repressor for homeodomain transcription factors. Alternative splicing results in four transcript variants encoding four distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIPK1/HIPK2/HIPK3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HIPK3
Gene Accession No. A4L9P5, O88850, O88904, Q86Z02, Q9ERH7, Q9H2X6, Q9H422, Q9QZR5
Gene Alias 1110014O20Rik; 1110062K04Rik; androgen receptor-interacting nuclear protein kinase; ANPK; B230339E18Rik; DYRK6; EMBL:ABO47653.1}; fas-interacting serine/threonine-protein kinase; FIST; FIST3; HCMV-interacting protein; hHIPk2; Hipk1; hipk1 {ECO:0000312; HIPK2; HIPK3; homeodomain interacting protein kinase 1; homeodomain interacting protein kinase 1-like protein; homeodomain interacting protein kinase 2; homeodomain interacting protein kinase 3; homeodomain-interacting protein kinase 1; Homeodomain-interacting protein kinase 2; homeodomain-interacting protein kinase 3; Homolog of protein kinase YAK1; KIAA0630; mir-1902; Myak; Nbak1; Nbak2; nbak3; nuclear body associated kinase 2b; nuclear body-associated kinase 1; Nuclear body-associated kinase 2; nuclear body-associated kinase 3; nuclear serine/threonine protein kinase; PKY; PRO0593; protein kinase Myak; Sialophorin tail-associated nuclear serine/threonine-protein kinase; Stank; YAK1
Gene Symbols HIPK1, HIPK2, HIPK3
Host Species Rabbit
Immunogen A synthesized peptide derived from human HIPK1(Accession Q86Z02), corresponding to amino acid residues Y331-V381.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10114, 15257, 15258, 15259, 204851, 28996, 362342, 365895, 83617
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.