Learn More
Promega HIPK1 Kinase Enzyme System, 10microg
Catalog No. PRV4066 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4066 10 μg
1 options
Catalog No. PRV4066 Supplier Promega Supplier No. V4066
Recombinant human HIPK1 (156-555) was expressed in Sf9 cells using an N-terminal GST tag. HIPK1 or homeodomain-interacting protein kinase 1 is a ser/thr protein kinase and a member of the HIPK family.
  • Easily Screen and Profile HIPK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C