Learn More
Promega HIPK1 Kinase Enzyme System, 1mg
Catalog No. PRV6396 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6396 1 mg
1 options
Catalog No. PRV6396 Supplier Promega Supplier No. V6396
Add to Cart
Add to Cart
Request a Quote
Recombinant human HIPK1 (156-555) was expressed in Sf9 cells using an N-terminal GST tag. HIPK1 or homeodomain-interacting protein kinase 1 is a ser/thr protein kinase and a member of the HIPK family.
  • Easily Screen and Profile HIPK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C