Learn More
Invitrogen™ HIPK1 Monoclonal Antibody (OTI3C4)

Description
HIPK1 Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | HIPK1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI3C4 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | HIPK1 |
| Gene Accession No. | Q86Z02 |
| Gene Alias | 1110062K04Rik; EMBL:ABO47653.1}; Hipk1; hipk1 {ECO:0000312; homeodomain interacting protein kinase 1; homeodomain interacting protein kinase 1-like protein; homeodomain-interacting protein kinase 1; KIAA0630; MYAK; NBAK2; nuclear body associated kinase 2b; nuclear body-associated kinase 2; Protein kinase Myak |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.