Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIPK1 Monoclonal Antibody (OTI3C4), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA807264

Description
The protein encoded by this gene belongs to the Ser/Thr family of protein kinases and HIPK subfamily. It phosphorylates homeodomain transcription factors and may also function as a co-repressor for homeodomain transcription factors. Alternative splicing results in four transcript variants encoding four distinct isoforms.Specifications
HIPK1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q86Z02 | |
HIPK1 | |
Human recombit protein fragment corresponding to amino acids 439-697 of human HIPK1 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI3C4 | |
Unconjugated | |
HIPK1 | |
Myak, Nbak2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
204851 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction