Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIPK1 Monoclonal Antibody (OTI5F1)
GREENER_CHOICE

Catalog No. PIMA526853
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526853 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526853 Supplier Invitrogen™ Supplier No. MA526853
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HIPK1 Monoclonal Antibody for Western Blot, IHC (P)

HIPK1 phosphorylates homeodomain transcription factors and may also function as a co-repressor for homeodomain transcription factors. The protein encoded by HIPK1 belongs to the Ser/Thr family of protein kinases and HIPK subfamily. Alternative splicing results in four transcript variants encoding four distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIPK1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI5F1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HIPK1
Gene Accession No. Q86Z02
Gene Alias 1110062K04Rik; EMBL:ABO47653.1}; Hipk1; hipk1 {ECO:0000312; homeodomain interacting protein kinase 1; homeodomain interacting protein kinase 1-like protein; homeodomain-interacting protein kinase 1; KIAA0630; MYAK; NBAK2; nuclear body associated kinase 2b; nuclear body-associated kinase 2; Protein kinase Myak
Gene Symbols HIPK1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 439-697 of HIPK1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 204851
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.