Learn More
HIPK1 Mouse anti-Human, Clone: OTI3C11, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF807258

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene belongs to the Ser/Thr family of protein kinases and HIPK subfamily. It phosphorylates homeodomain transcription factors and may also function as a co-repressor for homeodomain transcription factors. Alternative splicing results in four transcript variants encoding four distinct isoforms.Specifications
HIPK1 | |
Monoclonal | |
Unconjugated | |
HIPK1 | |
Myak, Nbak2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
204851 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI3C11 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q86Z02 | |
HIPK1 | |
Human recombit protein fragment corresponding to amino acids 439-697 of human HIPK1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.