Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HIPK1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115358
Description
Antibody detects endogenous levels of total HIPK1.
HIPK1 phosphorylates homeodomain transcription factors and may also function as a co-repressor for homeodomain transcription factors. The protein encoded by HIPK1 belongs to the Ser/Thr family of protein kinases and HIPK subfamily. Alternative splicing results in four transcript variants encoding four distinct isoforms.
Specifications
HIPK1 | |
Polyclonal | |
Unconjugated | |
HIPK1 | |
1110062K04Rik; EMBL:ABO47653.1}; Hipk1; hipk1 {ECO:0000312; homeodomain interacting protein kinase 1; homeodomain interacting protein kinase 1-like protein; homeodomain-interacting protein kinase 1; KIAA0630; MYAK; NBAK2; nuclear body associated kinase 2b; nuclear body-associated kinase 2; Protein kinase Myak | |
Rabbit | |
Affinity Chromatography | |
RUO | |
15257, 204851, 365895 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
A4L9P5, O88904, Q86Z02 | |
HIPK1 | |
A synthesized peptide derived from human HIPK1(Accession Q86Z02), corresponding to amino acid residues S9-S59. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction