Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIPK2 Monoclonal Antibody (3E9), Active Motif™
SDP

Catalog No. 501992775
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2775 10 μg
50-199-2774 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2775 Supplier Active Motif Supplier No. 39678
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HIPK2 is a conserved serine/threonine nuclear kinase that interacts with homeodomain transcription factors.

Specifications

Antigen HIPK2
Applications ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 3E9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene HIPK2
Gene Accession No. Q9H2X6
Gene Alias 1110014O20Rik; B230339E18Rik; BOS_4755; cb1009; hHIPk2; HIPK2; homeodomain interacting protein kinase 2; Homeodomain-interacting protein kinase 2; Nbak1; Nuclear body-associated kinase 1; PRO0593; si:dkey-16j24.2; si:dkey-239k7.1; Sialophorin tail-associated nuclear serine/threonine-protein kinase; Stank; wu:fk56d12
Gene Symbols HIPK2
Host Species Mouse
Immunogen This HIPk2 antibody was raised against a recombinant protein including amino acids 898-1051 of mouse HIPk2.
Purification Method Protein G
Quantity 10 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28996
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.