Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIPK3 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP224521APC
Description
HIPK3 Polyclonal antibody specifically detects HIPK3 in Human, Mouse, Rat, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
HIPK3 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
Androgen receptor-interacting nuclear protein kinase, DYRK6Fas-interacting serine/threonine-protein kinase, EC 2.7.11.1, FIST, FIST3ANPK, homeodomain interacting protein kinase 3, homeodomain-interacting protein kinase 3, PKYHomolog of protein kinase YAK1, YAK1 | |
A portion of amino acids 850-900 of human HIPK3 was used as the immunogen. | |
0.1 mL | |
Protein Kinase | |
10114 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
APC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Canine, Primate | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction