Learn More
Promega HIPK3 Kinase Enzyme System, 10microg
Catalog No. PRV4164 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4164 10 μg
1 options
Catalog No. PRV4164 Supplier Promega Supplier No. V4164
Recombinant human HIPK3 (163-562) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. HIPK3 (homeodomain interacting protein kinase 3) is a member of the HIPK family and is involved in apoptosis.
  • Easily Screen and Profile HIPK3 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C