Learn More
Invitrogen™ HIPK3 Polyclonal Antibody

Description
Immunogen sequence homology: Equine (83%).
Specifications
Specifications
| Antigen | HIPK3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene | HIPK3 |
| Gene Accession No. | O88850, Q9ERH7, Q9H422 |
| Gene Alias | Androgen receptor-interacting nuclear protein kinase; ANPK; DYRK6; Fas-interacting serine/threonine-protein kinase; FIST; FIST3; HCMV-interacting protein; HIPK3; homeodomain interacting protein kinase 3; homeodomain-interacting protein kinase 3; Homolog of protein kinase YAK1; mir-1902; nbak3; Nuclear body-associated kinase 3; nuclear serine/threonine protein kinase; PKY; RP1-8L15.1; YAK1 |
| Gene Symbols | HIPK3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.