Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIPK4 Monoclonal Antibody (292CT1.4.1)
GREENER_CHOICE

Catalog No. PIMA537528
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537528 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537528 Supplier Invitrogen™ Supplier No. MA537528
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HIPK4 Monoclonal Antibody for Western Blot

HIPK4 is a homeodomain-interacting protein kinase and has been demonstrated to phosphorylate p53. HIPK4 encodes a member of the homeodomain interacting protein kinase (HIPK) family of proteins. While other members of this family are found throughout vertebrates, this member is present only in mammals. Compared to other members of this family, the encoded protein lacks a nuclear localization signal and a C-terminal autoinhibitory domain. The encoded protein exhibits kinase activity and may phosphorylate the tumor suppressor protein p53.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIPK4
Applications Western Blot
Classification Monoclonal
Clone 292CT1.4.1
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene HIPK4
Gene Accession No. Q8NE63
Gene Alias Gm162; Hipk4; homeodomain interacting protein kinase 4; homeodomain-interacting protein kinase 4; LOW QUALITY PROTEIN: homeodomain-interacting protein kinase 4; testicular tissue protein Li 87
Gene Symbols HIPK4
Host Species Mouse
Immunogen HIPK4 recombinant protein.
Purification Method Euglobulin precipitation
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147746
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgM κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.