Learn More
Invitrogen™ HIPK4 Monoclonal Antibody (2E8)
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA523081
Catalog No. PIMA523081
Only null left
Add to Cart
Description
Description
Peptide Sequence: RGSSWEEGEH LGASAEPLAI LQRDEDGPNI DNMTMEAERP DPELFDPSSC PGEWLSEPDC TLESVRGPRA QGLPPRRSHQ HGPPRGATSF LQHVTGHH.
HIPK4 is a homeodomain-interacting protein kinase and has been demonstrated to phosphorylate p53. HIPK4 encodes a member of the homeodomain interacting protein kinase (HIPK) family of proteins. While other members of this family are found throughout vertebrates, this member is present only in mammals. Compared to other members of this family, the encoded protein lacks a nuclear localization signal and a C-terminal autoinhibitory domain. The encoded protein exhibits kinase activity and may phosphorylate the tumor suppressor protein p53.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HIPK4
Monoclonal
1 mg/mL
PBS with no preservative; pH 7.4
Q8NE63
HIPK4
HIPK4 (AAH34501.1, 519 a.a. ~ 616 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2a κ
ELISA, Western Blot
2E8
Unconjugated
HIPK4
Gm162; Hipk4; homeodomain interacting protein kinase 4; homeodomain-interacting protein kinase 4; LOW QUALITY PROTEIN: homeodomain-interacting protein kinase 4; testicular tissue protein Li 87
Mouse
Affinity chromatography
RUO
147746
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.