Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIPK4 Rabbit anti-Human, Mouse, Non-human Primate, Rat, Polyclonal, FabGennix

Catalog No. 502207339
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-7339 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-7339 Supplier Fabgennix Inc Supplier No. HIPK4401AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HIPK4 Polyclonal antibody specifically detects HIPK4 in Human, Mouse, Non-human Primate, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen HIPK4
Applications ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene HIPK4
Gene Accession No. Q3V016, Q4V793, Q8NE63
Gene Alias Gm162; Hipk4; homeodomain interacting protein kinase 4; homeodomain-interacting protein kinase 4; LOW QUALITY PROTEIN: homeodomain-interacting protein kinase 4; testicular tissue protein Li 87
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 470-550 on human Homeodomain-interacting protein kinase 4.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147746, 233020, 308449, 468883
Target Species Human, Mouse, Non-human Primate, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.