Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIPPI Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519110
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519110 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519110 Supplier Invitrogen™ Supplier No. PA519110
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.

HIPPI is required for the formation of cilia. Plays an indirect role in sonic hedgehog signaling, cilia being required for all activity of the hedgehog pathway. Has pro-apoptotic function via its interaction with HIP1, leading to recruit caspase-8 (CASP8) and trigger apoptosis. Has the ability to bind DNA sequence motif 5'-AAAGACATG-3' present in the promoter of caspase genes such as CASP1, CASP8 and CASP10, suggesting that it may act as a transcription regulator; however the relevance of such function remains unclear.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIPPI
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene IFT57
Gene Accession No. Q8BXG3, Q9NWB7
Gene Alias 4833420A15Rik; dermal papilla-derived protein 8; DERP8; Esrrbl1; estrogen-related receptor beta like 1; estrogen-related receptor beta-like protein 1; FLJ10147; HIP1 protein interactor; HIP1-interacting protein; Hippi; huntingtin interacting protein-1 interacting protein; huntingtin-interacting protein-1 protein interactor; IFT57; intraflagellar transport 57; intraflagellar transport 57 homolog; intraflagellar transport protein 57 homolog; MHS4R2; Vestrogen-related receptor beta like 1
Gene Symbols IFT57
Host Species Goat
Immunogen Synthetic peptide sequence (EQPQEYDDPNATISN ) corresponding to the internal amino acids of IFT57 (aa 103-117).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55081, 73916
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.