Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hippocalcin Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306199PE
Description
Hippocalcin Polyclonal antibody specifically detects Hippocalcin in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Hippocalcin | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| BDR2, Calcium-binding protein BDR-2, hippocalcin, neuron specific calcium-binding protein hippocalcin, neuron-specific calcium-binding protein hippocalcin | |
| Produced in rabbits immunized with E. coli-derived Human Hippocalcin fragment. | |
| 0.1 mL | |
| Neuroscience | |
| 3208 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| PE | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction