Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hippocalcin like 4 Antibody (OTI3C3), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271923R
Description
Hippocalcin like 4 Monoclonal specifically detects Hippocalcin like 4 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Hippocalcin like 4 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
HPCAL4 | |
Recombinant protein fragment corresponding to amino acids 2-191 of human Hippocalcin like 4 (NP_057341) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
OTI3C3 | |
Western Blot | |
hippocalcin like 4, hippocalcin-like protein 4, HLP4DKFZp761G122 | |
Mouse | |
Affinity Purified | |
Vision | |
51440.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction