Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hippocalcin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560290
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560290 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560290 Supplier Invitrogen™ Supplier No. PA560290
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MVSSVMKMPE DESTPEKRTE KIFRQMDT Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

The protein encoded by this gene is a member of neuron-specific calcium-binding proteins family found in the retina and brain. This protein is associated with the plasma membrane. It has similarities to proteins located in the photoreceptor cells that regulate photosignal transduction in a calcium-sensitive manner. This protein displays recoverin activity and a calcium-dependent inhibition of rhodopsin kinase. It is identical to the rat and mouse hippocalcin proteins and thought to play an important role in neurons of the central nervous system in a number of species.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hippocalcin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HPCA
Gene Accession No. P84074
Gene Alias BDR2; calcium-binding protein; calcium-binding protein BDR-2; DYT2; hippocalcin; Hpca; hypothetical protein MGC73126; neuron-specific calcium-binding protein hippocalcin; P23K; zgc:73126
Gene Symbols HPCA
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Hippocalcin. Recombinant protein control fragment (Product #RP-104356).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3208
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.