Learn More
Invitrogen™ Hippocalcin Recombinant Rabbit Monoclonal Antibody (6N2A6)

Description
Positive Samples: PC-12, MCF7, C6, Mouse testis, Rat brain Immunogen sequence: MGKQNSKLRP EMLQDLRENT EFSELELQEW YKGFLKDCPT GILNVDEFKK IYANFFPYGD ASKFAEHVFR TFDTNSDGTI DFREFIIALS VTSRGRLEQK.
Specifications
Specifications
| Antigen | Hippocalcin |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 6N2A6 |
| Concentration | 0.63 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | HPCA |
| Gene Accession No. | P84074, P84075, P84076 |
| Gene Alias | BDR2; calcium-binding protein; calcium-binding protein BDR-2; DYT2; hippocalcin; Hpca; hypothetical protein MGC73126; neuron-specific calcium-binding protein hippocalcin; P23K; zgc:73126 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.