Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Detergent Removal Spin Columns & Plates

Catalog No. pi88306
Click to view available options
Quantity:
10 mL
2 x 96-well Plates
24 x 0.1 mL Columns
25 x 125 μL Columns
25 x 500 μL Columns
5 mL
5 x 2 mL Columns
5 x 4 mL Columns
Product Line:
HiPPR™
Pierce™
Product Type:
Detergent Removal Resin
Spin Column
Spin Plate

Rapidly eliminate high concentrations of detergents with minimal sample loss with detergent removal resins. Thermo Scientific HiPPR (High Protein and Peptide Recovery) resin is recommended for protein concentrations up to 100 μg/mL, while Thermo Scientific Pierce resin is suitable for concentrations over 100 μg/mL.

Detergents and surfactants used to prepare protein and peptide samples can interfere with analysis by ELISA, isoelectric focusing, and mass spectrometry (MS). Removing detergents from peptide samples is especially challenging and critical for MS analysis. Even low concentrations of detergents will contaminate instruments and interfere with column binding, elution, and peptide ionization. Thermo Scientific HiPPR and Pierce detergent removal resins are ideal for rapid removal of common detergents from tryptic digests to improve the results of LC-MS/MS and MALDI-MS analysis.

Optimized—removes >95% of detergent from samples of proteins or peptides
Fast—sample processing takes less than 15 minutes
Effective—eliminates detergent-interference in downstream applications like ELISA, isoelectric focusing, and mass spectrometry

Pierce Detergent Removal Resin
Pierce Detergent Removal Resin efficiently binds to and removes high concentrations of detergents from 10 μL to 1 mL samples with minimal sample loss. These microcentrifuge columns come prepacked with various volumes of proprietary resin and are best for processing samples with proteins or peptide concentrations greater than 100 μg/mL.

HiPPR Detergent Removal Spin Column Kit
HiPPR Detergent Removal resin has been optimized for sample concentrations of 1 to 100 μg/mL and is ideal for removing commonly used detergents, including SDS, Triton™ X-100, NP-40 and CHAPS at concentrations of 0.5-1%. The resin is available in pre-filled spin columns, 96-well filter spin plates, and as loose resin for customization.

Available formats:
Resin slurries—bulk resin supplied with empty 0.8-mL spin columns
Spin columns—0.1 mL micro-centrifuge columns
96-well spin plates—pre-dispensed 96-well filter plates, compatible with centrifugation and vacuum manifold systems for manual or automated purification

TRUSTED_SUSTAINABILITY

Specifications

Description Microcentrifuge, Spin Column
Product Type Spin Column
Content And Storage Upon receipt store at 4°C.
Column Type Proprietary Resin, Affinity
Stationary Phase Proprietary Ligand
Shipping Condition Ambient
Product Line HiPPR™
Purification Target Detergent Removal
Quantity 24 x 0.1 mL Columns
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.