Learn More
Thermo Scientific™ HiPPR™ Detergent Removal Spin Column Kit, 5 mL PROMO
Description
- Optimized—removes >95% of detergent from samples of proteins or peptides
- Fast—sample processing takes less than 15 minutes
- Effective—eliminates detergent-interference in downstream applications like ELISA, isoelectric focusing, and mass spectrometry
Pierce Detergent Removal Resin
Pierce Detergent Removal Resin efficiently binds to and removes high concentrations of detergents from 10 μL to 1 mL samples with minimal sample loss. These microcentrifuge columns come prepacked with various volumes of proprietary resin and are best for processing samples with proteins or peptide concentrations greater than 100 μg/mL.
HiPPR Detergent Removal Spin Column Kit
HiPPR Detergent Removal resin has been optimized for sample concentrations of 1 to 100 μg/mL and is ideal for removing commonly used detergents, including SDS, Triton™ X-100, NP-40 and CHAPS at concentrations of 0.5-1%. The resin is available in pre-filled spin columns, 96-well filter spin plates, and as loose resin for customization.
Available formats:
- Resin slurries—bulk resin supplied with empty 0.8 mL spin columns
- Spin columns—0.1 mL micro-centrifuge columns
- 96-well spin plates—pre-dispensed 96-well filter plates, compatible with centrifugation and vacuum manifold systems for manual or automated purification
Specifications
Specifications
| Product Type | Spin Column |
| Content And Storage | Detergent Removal Resin, 5 mL, Spin Columns Accessory Pack, 54 Columns, Store at 4°C. |
| Column Type | Proprietary Resin, Affinity |
| Stationary Phase | Proprietary Ligand |
| Product Line | HiPPR |
| Purification Target | Detergent Removal |
| Sufficient For | 54 Samples |
| Quantity | 5 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.