Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hippuric acid, 98%

Catalog No. AAA126900B
Change view
Click to view available options
Quantity:
100 g
500 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA126900B 1000 g
AAA1269022 100 g
AAA1269036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA126900B Supplier Thermo Scientific Chemicals Supplier No. A126900B
Add to Cart

CAS: 495-69-2 | C9H9NO3 | 179.18 g/mol

Hippuric acid is used as a intermediate for the manufacturing medicine and other organic compounds. Hippuric acid can be used to study cell biology, chemical biology, bioactive small molecules, amino acid derivatives, peptide synthesis, chemical synthesis and nutrition. Hippuric acid has been used to inform the metabolism and urinary excretion of procyanidins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hippuric acid is used as a intermediate for the manufacturing medicine and other organic compounds. Hippuric acid can be used to study cell biology, chemical biology, bioactive small molecules, amino acid derivatives, peptide synthesis, chemical synthesis and nutrition. Hippuric acid has been used to inform the metabolism and urinary excretion of procyanidins.

Solubility
Soluble in water.

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 495-69-2
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.18
MDL Number MFCD00002692
InChI Key QIAFMBKCNZACKA-UHFFFAOYSA-N
Synonym hippuric acid, n-benzoylglycine, glycine, n-benzoyl, benzoylglycine, benzamidoacetic acid, benzoylaminoacetic acid, hippurate, hippursaeure, phenylcarbonylaminoacetic acid, benzamidoessigsaeure
PubChem CID 464
ChEBI CHEBI:18089
IUPAC Name 2-benzamidoacetic acid
SMILES OC(=O)CNC(=O)C1=CC=CC=C1

Specifications

Melting Point 188°C to 191°C
Density 1.371
Linear Formula C6H5CONHCH2CO2H
Quantity 1000 g
Beilstein 1073987
Merck Index 14,4716
Solubility Information Soluble in water.
Formula Weight 179.18
Percent Purity 98%
Chemical Name or Material Hippuric acid
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
EINECSNumber 207-806-3
RTECSNumber MR8150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.