Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiPrep™ HIC FF 16/10 Columns

Catalog No. 45002077 Shop All Cytiva Products
Change view
Click to view available options
Product Type:
Butyl Sepharose
Octyl Sepharose
Phenyl Sepharose (high sub)
Support Type:
4% Cross-linked Agarose
6% Cross-linked Agarose
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Support Type
45-002-077 Butyl Sepharose 4% Cross-linked Agarose
45-002-076 Octyl Sepharose 4% Cross-linked Agarose
45-002-079 Phenyl Sepharose (high sub) 6% Cross-linked Agarose
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 45-002-077 Supplier Cytiva Supplier No. 28936547
Add to Cart
Add to Cart

Produce fast, preparative separation of proteins and other biomolecules. Cytiva Life Sciences™ HiPrep™ HIC FF 16/10 Columns are high capacity with excellent flow properties, and reliable, reproducible separations.

The 20mL HIC columns provide fast, preparative separation of proteins and other biomolecules. They are ready-to-use with four media choices: Phenyl Sepharose 6 Fast Flow (high sub); Phenyl Sepharose 6 Fast Flow (low sub); Butyl Sepharose 4 Fast Flow; and Octyl Sepharose 4 Fast Flow.

Features:

  • Offer high capacity, excellent flow properties and reliable, reproducible separations
  • Compatible with simple pump-based configurations or chromatography systems (e.g., AKTA)

Specifications (H2O at room temp.)

  • Dimensions: 1.6 x 10cm
  • Recommended flowrate: 2-10mL/min. (60-300cm/h)
  • Maximum flowrate: 10mL/min. (300cm/hr.)
  • Max. pressure over packed bed: 1.5bar (22psi, 0.15MPa)
  • Column hardware pressure limit: 5bar (73psi, 0.5MPa)
  • Storage: 20% ethanol
  • Storage temperature: 4° to 30°C

TRUSTED_SUSTAINABILITY

Specifications

Buffer 1M NaOH, 30% Isopropanol, 70% Ethanol, 6M Guanidine Hydrochloride, 30% Aceto nitrile, 1mM HCl
Product Type Butyl Sepharose
Bed Volume 20mL
Diameter (English) Inner 0.62 in.
Diameter (Metric) Inner 16mm
Quantity 1 x 20 mL
Height (English) 3.93 in.
Height (Metric) 100mm
Flow Rate >300 cm/hr.
Material Polypropylene
Ligand Type Butyl
Max. Pressure 5 bar
pH Range 3 to 13
Particle Size 45 to 165μm
Support Type 4% Cross-linked Agarose
Content And Storage 4° to 30°C
Column Type Ion Exchange Column
For Use With (Application) Provide fast, preparative separation of Proteins and other biomolecules by hydrophobic interaction chromato graphy (HIC), initial and intermediate purification, removing bulk impurities
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.