Learn More
Description
- Long-term chemical and physical stability from hydrophilic, rigid allyl dextran/bisacrylamide matrix
- Recommended linear flowrate 5 to 15cm/hr.
Designed for separating peptides and small proteins
Specifications
Specifications
| Quantity | Each |
| Flow Rate | <0.5 mL/min. |
| Particle Size | 25 to 75 μm |
| For Use With (Equipment) | Low to medium pressure AKTA™ systems |
| Bed Volume | 120 mL |
| Product Type | Sephacryl HR Column |
| Diameter (Metric) Inner | 1.6cm |
| For Use With (Application) | Separation of biomolecules with maximum recovery and minimum nonspecific adsorption with long term chemical and physical stability resulting from the hydrophilic, rigid Allyl Dextran/Bisacrylamide matrix |
| Height (English) | 23.62 in. |
| Height (Metric) | 60cm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.