Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ HiPur™ Denatured Ethyl Alcohol

Catalog No. p-4532368 Shop All Epredia Products
Click to view available options
Quantity:
1 gal.
5 gal.
This item is not returnable. View return policy

Catalog No. 22-110-880

Supplier: Epredia™ 92001

Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Choose the option that best fits your work, HiPur™ Denatured Ethanol is available in one or five gallon sizes.

  • Maximum of 0.1% water
  • Maximum residue content of 0.003%
  • Anhydrous formula is denatured with Methanol, MIBK, Ethyl Acetate and Heptane

Specifications

Product Type Alcohol
Quantity 1 gal.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.