Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIRA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562234
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562234 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562234 Supplier Invitrogen™ Supplier No. PA562234
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LKLPIPSPQR AFTLQVSSDP SMYIEVENEV TVVGGVKLSR LKCNREGKEW ETVLTSRILT AAGSCDVVCV ACEKRMLSVF STCGRRLLSP ILLPSPISTL HC Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.

HIRA is a histone chaperone that preferentially places the variant histone H3.3 in nucleosomes. Orthologs of this protein in yeast, flies, and plants are necessary for the formation of transcriptionally silent heterochromatin. It plays an important role in the formation of the senescence-associated heterochromatin foci. These foci likely mediate the irreversible cell cycle changes that occur in senescent cells. It is considered the primary candidate protein in some haploinsufficiency syndromes such as DiGeorge syndrome, and insufficient production of the gene may disrupt normal embryonic development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIRA
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HIRA
Gene Accession No. P54198
Gene Alias AA138857; D16Ertd95e; DGCR1; DiGeorge critical region gene 1; Gm15797; HIR; HIR histone cell cycle regulation defective homolog A; Hira; histone cell cycle regulation defective homolog A; histone cell cycle regulation defective homolog A (S. cerevisiae); histone cell cycle regulator; N28177; OTTMUSG00000026264; Protein HIRA; TUP1; TUP1-like enhancer of split gene 1; TUP1-like enhancer of split protein 1; TUPLE1
Gene Symbols HIRA
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HIRA. Recombinant protein control fragment (Product #RP-103085).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7290
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.