Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIRA Rabbit anti-Human, Mouse, Polyclonal, liquid, OriGene

Rabbit Polyclonal Antibody
Supplier: OriGene TA890126

Description
HIRA is a histone chaperone that preferentially places the variant histone H3.3 in nucleosomes. Orthologs of this protein in yeast, flies, and plants are necessary for the formation of transcriptionally silent heterochomatin. It plays an important role in the formation of the senescence-associated heterochromatin foci. These foci likely mediate the irreversible cell cycle changes that occur in senescent cells. It is considered the primary candidate protein in some haploinsufficiency syndromes such as DiGeorge syndrome, and insufficient production of the gene may disrupt normal embryonic development.Specifications
HIRA | |
Polyclonal | |
PBS with 50% glycerol and 0.02% sodium azide | |
P54198, Q61666 | |
HIRA | |
Recombit protein of human HIRA | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
HIRA | |
DGCR1, TUP1, TUPLE1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15260, 7290 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction