Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIRA Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$149.90 - $370.72
Specifications
| Antigen | HIRA |
|---|---|
| Dilution | Western Blot 1:1000, ELISA 1:500-1:200 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
HIRA Polyclonal antibody specifically detects HIRA in Mouse samples. It is validated for Western Blot, ELISASpecifications
| HIRA | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Chromatin Modifiers, Stem Cells | |
| Whole antiserum | |
| DGCR1DiGeorge critical region gene 1, HIR, HIR (histone cell cycle regulation defective) homolog A (S. cerevisiae), HIR histone cell cycle regulation defective homolog A (S. cerevisiae), TUP1, TUP1-like enhancer of split protein 1, TUPLE1protein HIRA | |
| Polyclonal antibody raised in rabbit against mouse HIRA (histone cell cycle regulation defective homolog A), using a KLH-conjugated synthetic peptide containing an amino acid sequence from the central part of the protein (1). | |
| Whole Antiserum |
| Western Blot 1:1000, ELISA 1:500-1:200 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Mouse | |
| P54198 | |
| 7290 | |
| Primary | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title