Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HIRA Recombinant Rabbit Monoclonal Antibody (6Q7Y6)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542899
This item is not returnable.
View return policy
Description
Positive test controls include: HeLa, 293T. Immunogen sequence: PSVLTTPSKI EPMKAFDSRF TERSKATPGA PALTSMTPTA VERLKEQNLV KELRPRDLLE SSSDSDEKVP LAKASSLSKR KLELEVETVE KKKKGRPRKD SRLMPVSLSV QSPAALTAEK EAMCLS.
HIRA is a histone chaperone that preferentially places the variant histone H3.3 in nucleosomes. Orthologs of this protein in yeast, flies, and plants are necessary for the formation of transcriptionally silent heterochromatin. It plays an important role in the formation of the senescence-associated heterochromatin foci. These foci likely mediate the irreversible cell cycle changes that occur in senescent cells. It is considered the primary candidate protein in some haploinsufficiency syndromes such as DiGeorge syndrome, and insufficient production of the gene may disrupt normal embryonic development.
Specifications
HIRA | |
Recombinant Monoclonal | |
1.5 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P54198 | |
HIRA | |
Recombinant fusion protein containing a sequence corresponding to amino acids 550-675 of human HIRA (P54198). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Western Blot | |
6Q7Y6 | |
Unconjugated | |
HIRA | |
AA138857; D16Ertd95e; DGCR1; DiGeorge critical region gene 1; Gm15797; HIR; HIR histone cell cycle regulation defective homolog A; Hira; histone cell cycle regulation defective homolog A; histone cell cycle regulation defective homolog A (S. cerevisiae); histone cell cycle regulator; N28177; OTTMUSG00000026264; Protein HIRA; TUP1; TUP1-like enhancer of split gene 1; TUP1-like enhancer of split protein 1; TUPLE1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
7290 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction