Learn More
Sigma Aldrich HIRMA PMMA 3D Printing Filament
Catalog No. 503125893 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
50-312-5893 1 Ea.
1 options
Catalog No. 50-312-5893 Supplier Sigma Aldrich Supplier No. TRD3D00381EA
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Parameters:Extruder (nozzle) temperature: 230 +/-10°CBed temperature: 100 +/-10°CPrint speed: 30-50 mm/sFan: 0-20 %Minimum nozzle diameter: 0.3 mmPlate materials: kapton tape with glue, blue tape with glue, etc.
Specifications
Quantity 1 Ea.