Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

His Tag Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC0501R100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC0501R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC0501R100 Supplier R&D Systems Supplier No. IC0501R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

His Tag Monoclonal specifically detects His Tag in Multi-Species samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen His Tag
Applications Flow Cytometry
Classification Monoclonal
Clone AD1.1.10R
Conjugate Alexa Fluor 647
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Alias 6 His epitope tag, 6X His, 6X-His, H, HHHHHH epitope tag, HHHHHH tag, HIS, His tag, polyHistidine, Poly-histidine
Host Species Mouse
Immunogen His-tagged peptide
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects proteins containing accessible consecutive histidine regions. The antibody detects His tags located at the amino- or carboxyl- terminus.
Target Species Multi-species
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.