Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
His Tag Antibody (3D5), Alexa Fluor™ 647, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP281023AF647
Description
His Tag Monoclonal antibody specifically detects His Tag in Epitope Tag samples. It is validated for Western Blot, ELISA.Specifications
His Tag | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rat | |
Protein A purified | |
RUO | |
Primary | |
Store at 4°C in the dark. | |
IgG2a κ |
Western Blot, ELISA | |
3D5 | |
Western Blot, ELISA | |
6 His epitope tag, 6X His, 6X-His, H, HHHHHH epitope tag, HHHHHH tag, HIS, His tag, Poly-histidine | |
Purified his tagged Fab fragment. | |
0.1 mL | |
Cellular Markers, Epitope Tags | |
This antibody binds to carboxy-terminal oligo-histidine tags (6x His tags) of fusion proteins. It also has affinity for internal His-tags but no affinity for N-terminal His-tags. | |
Purified | |
Epitope Tag |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction