Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
His Tag Antibody (AD1.1.10), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB050X
Description
His Tag Monoclonal antibody specifically detects His Tag in Broad Species Reactivity samples. It is validated for Western Blot, Western Blot, Flow Cytometry, Affinity Purification, CyTOFSpecifications
His Tag | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Flow Cytometry, Affinity Purification, CyTOF | |
AD1.1.10 | |
Western Blot, Simple Western, Flow Cytometry, Immunoaffinity Purification, CyTOF-ready | |
6 His epitope tag, 6-His Tag, 6X His, 6X His Tag, 6X-His, 6x-His Tag, H, Hexa His tag, HHHHHH epitope tag, HHHHHH tag, HIS, His tag, His6-Tag, polyHistidine, Poly-histidine, polyhistidine tag | |
His-tagged peptide | |
0.1 mL | |
Cellular Markers, Epitope Tags | |
Broad Species Reactivity | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction