Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
His Tag Antibody (HIS.H8), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP231055JF549
Description
His Tag Monoclonal antibody specifically detects His Tag in Epitope Tag samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence.Specifications
HisTag | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein A purified | |
Cellular Markers, Epitope Tags | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
HIS.H8 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
6 His epitope tag, 6X His, 6X-His, H, HHHHHH epitope tag, HHHHHH tag, HIS, His tag, Poly-histidine | |
6x His synthetic peptide. | |
0.1 mL | |
Primary | |
Purified | |
Epitope Tag |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction