Description
- Control Kit
- 30 HisTag Check&Go! Strips
- 1 cryovial of 10X Running Buffer
- Requires 96-wells low binding plate (not supplied)
- Unlike traditional methods of assessing the success of your protein expression and purification, which can be tedious and time-consuming, His-Tag Check&Go! is a simple qualitative immunochromatography test that requires only 10–15 minutes of your time and very small quantities of your sample
- The key component of the kit is a nitrocellulose membrane containing a ‘Test line’ of immobilized His-Tag protein on the ‘Test line’ (T line) and a ULFA-tagged protein on the ‘Control line’ (C line)
- A 40nm InnovaCoat(R)-GOLD anti-His-Tag antibody and 40nm InnovaCoat(R)-GOLD anti-ULFA tag conjugate are used for detection
- This product is for research use only and is not approved for use in humans or in clinical diagnosis
- Kits are guaranteed for 6 months from date of receipt
Specifications
Specifications
| Kit Contents | 30 HisTag Check&Go! Strips and 1 cryovial of 10X Running Buffer |
| Quantity | 30 Tests |
| For Use With (Application) | Verify and monitor successful expression of His-tagged recombinant proteins in one easy step, prior to performing other downstream applications |