Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ His Tag Monoclonal Antibody

Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ 707963
Description
The His Tag™ Monoclonal Antibody is a mouse monoclonal antibody (IgG1) directed against the His Tag sequence encoded by many of the Novagen™ expression vectors as well as many other commercially available vectors. The antibody recognizes five consecutive histidine residues regardless of the surrounding amino acid context. The high affinity (Kd = 5 x 10-8 to 1 x 10-9 M) enables sensitive, specific detection of His Tag fusion proteins at antibody concentrations of 0.1-0.2μg/ml. The His Tag Monoclonal Antibody binds to N-terminal, C-terminal, and internal His Tag sequences. This antibody also detects the recombinant marker bands in the Perfect Protein™ and Trail Mix™ Western Markers for convenient visualization of accurate internal standards on Western blots*. For lowest background in Western blotting applications, Alkali-soluble Casein is recommended as a blocking agent. The 100μg package size provides enough purified antibody for 1000 ml of working solution.
Specifications
His Tag | |
Monoclonal | |
Lyophilized, BSA-free | |
100 μg | |
Primary | |
-20°C | |
IgG1 |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
RUO | |
HisHisHisHisHis, N-terminal, C-terminal or internal | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction