Learn More
MilliporeSigma™ His Tag Monoclonal Antibody
Mouse Monoclonal Antibody
Supplier:  MilliporeSigma™ 707964
 View more versions of this product
Catalog No. 70-796-4MI
Only null left
Add to Cart
Description
Description
The His Tag™ Monoclonal Antibody is a mouse monoclonal antibody (IgG1) directed against the His Tag sequence encoded by many of the Novagen™ expression vectors as well as many other commercially available vectors. The antibody recognizes five consecutive histidine residues regardless of the surrounding amino acid context. The high affinity (Kd = 5 x 10-8 to 1 x 10-9 M) enables sensitive, specific detection of His Tag fusion proteins at antibody concentrations of 0.1-0.2μg/ml. The His Tag Monoclonal Antibody binds to N-terminal, C-terminal, and internal His Tag sequences. This antibody also detects the recombinant marker bands in the Perfect Protein™ and Trail Mix™ Western Markers for convenient visualization of accurate internal standards on Western blots*. For lowest background in Western blotting applications, Alkali-soluble Casein is recommended as a blocking agent. The 100μg package size provides enough purified antibody for 1000 ml of working solution.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
His Tag
Monoclonal
Lyophilized, BSA-free
3 μg
Primary
-20°C
IgG1
Immunoprecipitation, Western Blot
Unconjugated
Mouse
RUO
HisHisHisHisHis, N-terminal, C-terminal or internal
Lyophilized
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only. Not for use in diagnostic procedures.