Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
His Tag Mouse anti-Nonspecies specific, DyLight 550, Clone: 33D10.D2, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP142781R0.2ML
Description
His Tag Monoclonal specifically detects His Tag in Epitope Tag samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Fluorophore-linked immunosorbent assay.Specifications
His Tag | |
Monoclonal | |
DyLight 550 | |
6 His epitope tag, 6X His, 6X-His, H, HHHHHH epitope tag, HHHHHH tag, HIS, His tag, Poly-histidine | |
His Tag antibody was produced in mice by repeated immunizations with 6X His epitope tag peptide H-H-H-H-H-H conjugated to KLH using maleimide. | |
0.2 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry | |
33D10.D2 | |
50 mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Non-species specific | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction