Learn More
His Tag Mouse, Clone: 6G2A9, THE™, GenScript™
Mouse Monoclonal Antibody
Supplier: Genscript Corporation A00186100

Description
Reconstitute the lyophilized antibody with deionized water (or equivalent) to a final concentration of 0.5 mg/mL. Monoclonal antibodies specific to six histidine tags can greatly improve the effectiveness of several different kinds of immunoassays, helping researchers identify, detect, and purify polyhistidine fusion proteins in bacteria, insect cells, and mammalian cells. However, since 6XHis-tag is poorly immunogenic, it needs to be conjugated to KLH or some other carrier as an immunogen. After hundreds of selection cycles, researchers at GenScript successfully isolated an antibody against His-tag. THE™ His Tag Antibody, mAb, Mouse (subtype IgG1) has very high affinity. Tests performed at GenScript show that the antibody can also recognize 4xHis- and 5xHis-tags. This means that even if the 6xHis-tag is only partially exposed, it will still be recognized and bound by this antibody. THE™ His Tag mAb is produced from mice ascites and purified by protein A affinity column. This antibody recognizes native as well as denatured forms of synthetic polyhistidine and polyhistidine-tagged fusion proteins. The product reacts with fusion proteins expressed in bacteria, insect cells, and mammalian cells. THE™ His Tag mAb recognizes His tags placed at N-terminal, C-terminal, and internal regions of fusion proteins. THE™ His Tag mAb can be used in Western blot analyses, Dot Blot analyses, ELISA, immunofluorescent staining, and flow cytometry of cultured cells.
His Tag Monoclonal antibody specifically detects His Tag in Tag samples. It is validated for ELISA, Flow Cytometry, Fluorescence Resonance Energy Transfer (FRET), Immunocytochemistry, Immunoprecipitation, Western BlotSpecifications
His Tag | |
Monoclonal | |
Unconjugated | |
6His tag; 6X His; His Tag; Histidine Tag | |
A synthetic peptide HHHHHH coupled to KLH | |
100 μg | |
Primary | |
Antibody | |
IgG1 κ |
ELISA, Flow Cytometry, Fluorescence Resonance Energy Transfer (FRET), Immunocytochemistry, Immunoprecipitation, Western Blot | |
6G2A9 | |
PBS with 0.02% sodium azide | |
Mouse | |
Protein A | |
RUO | |
-20°C or -80°C if preferred | |
Lyophilized | |
Tag |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.