Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

His-tag Rabbit, Biotin, Polyclonal, GenScript™

Catalog No. 50272495
Encompass_Preferred
Change view
Click to view available options
Quantity:
40 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-272-495 40 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-272-495 Supplier Genscript Corporation Supplier No. A0020540
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute the lyophilized powder with deionized water (or equivalent) to an antibody concentration of 0.5 mg/mL. GenScript His-tag antibody [Biotin], pAb, Rabbit is the biotin conjugated Rabbit anti His antibody. The antibody recognizes fusion protein with His tag which is located at either C-terminal, or internal regions of the protein. It is suitable for detecting the expression level of His tagged proteins.GenScript His-tag antibody [Biotin], pAb, Rabbit is the biotin conjugated Rabbit anti His antibody. It is suitable for detecting the expression level of His tagged proteins.

His-tag Polyclonal antibody specifically detects His-tag in Tag samples. It is validated for Western Blot

Specifications

Antigen His-tag
Applications Western Blot
Classification Polyclonal
Conjugate Biotin
Formulation PBS with 1% BSA and 0.02% sodium azide; pH 7.4
Gene Alias 6His tag; 6X His; His Tag; Histidine Tag
Host Species Rabbit
Immunogen His epitope tag peptide HHHHHH conjugated to KLH.
Purification Method Affinity chromatography
Quantity 40 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Lyophilized
Isotype IgG
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.