Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

His-Tag Rabbit, Clone: RM146 (Biotin), Abnova™

Catalog No. 89035316 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-316 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-316 Supplier Abnova Corporation Supplier No. MAB14964
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Rabbit monoclonal antibody raised against His-Tag.

  • High specificity
  • High reactivity

Specifications

Antigen His-Tag
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone RM146
Conjugate Biotin
Description Rabbit monoclonal antibody raised against His-Tag.
Dilution ELISA (0.01-0.5 ug/mL) Flow Cytometry (0.5-2 ug/mL) Immunocytochemistry (0.5-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.1-1 ug/mL) Immunoprecipitation Western Blotting (0.1-0.5 ug/mL) The optimal wor
Formulation In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
Host Species Rabbit
Immunogen A mixture of synthetic peptides corresponding to N-terminus region of 6X His-Tag and C-terminus region of 6X His-Tag.
Purification Method Protein A purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.